missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phleomycin, 20 mg/ml in 5mM HEPES buffer, sterile-filtered

Catalog No. AAJ67027XF
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67027XF 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67027XF Supplier Thermo Scientific Chemicals Supplier No. J67027XF

CAS: 11006-33-0 | C51H75N17O21S2 | 1326.379 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11006-33-0
Molecular Formula C51H75N17O21S2
Molecular Weight (g/mol) 1326.379
MDL Number MFCD00131846
InChI Key QRBLKGHRWFGINE-UGWAGOLRSA-N
PubChem CID 72511
IUPAC Name 2-[2-[2-[[2-[[4-[[2-[[6-amino-2-[3-amino-1-[(2,3-diamino-3-oxopropyl)amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[(2R,3S,4S,5S,6S)-3-[(2S,3R,4R,5S)-4-carbamoyl-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)-
SMILES CC1=C(N=C(N=C1N)C(CC(=O)N)NCC(C(=O)N)N)C(=O)NC(C(C2=CN=CN2)OC3C(C(C(C(O3)CO)O)O)(C)OC4C(C(C(C(O4)CO)O)(C(=O)N)O)O)C(=O)NC(C)C(C(C)C(=O)NC(C(=O)C)C(=O)NCCC5=NC(CS5)C6=NC(=CS6)C(=O)N)O

Specifications

Type Phleomycin
Physical Form Liquid
Packaging Plastic Bottle
Quantity 1 mL
Chemical Name or Material Phleomycin
Hazard Category H317-H351
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
RTECSNumber SY0525000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only