missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ PHKG2 Recombinant Human Protein
PHKG2 is a testis/liver isoform of the phosphorylase kinase gamma subunit.
$770.00 - $11260.00
Specifications
Accession Number | NP_000285 |
---|---|
Formulation | 50mM Tris (pH 7.5), 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 73.9kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV4555
|
Invitrogen™
PV4555 |
10μg |
Each for $770.00
|
|
|||||
PR7753A
|
Invitrogen™
PR7753A |
100μg |
Each for $2,655.00
|
|
|||||
PV4556
|
Invitrogen™
PV4556 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Specifications
NP_000285 | |
73.9kDa | |
RUO - research use only | |
PHK Kinase Family | |
Recombinant | |
Baculovirus |
50mM Tris (pH 7.5), 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
PHKG2 | |
Human | |
GST Tag |