Invitrogen™ PHKG2 Recombinant Human Protein DFS Item
PHKG2 is a testis/liver isoform of the phosphorylase kinase gamma subunit.
Manufacturer:  Invitrogen™ LSPV4556
 View more versions of this product
Catalog No. PV4556
Specifications
Specifications
NP_000285
Frozen
73.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
PHK Kinase Family
Yes
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
50mM Tris (pH 7.5), 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
PHKG2
Baculovirus
Human
GST Tag
Documents