Invitrogen™ PHKG2 Recombinant Human Protein DFS Item
PHKG2 is a testis/liver isoform of the phosphorylase kinase gamma subunit.
Manufacturer:  Invitrogen™ LSPV4555
 View more versions of this product
Catalog No. PV4555
Specifications
Specifications
NP_000285
50mM Tris (pH 7.5), 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
10μg
RUO - research use only
Full Length, Recombinant
PHKG2
Kinase Assay
Baculovirus
Human
GST Tag
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
73.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
N-Terminal
Serine/Threonine Kinases
phosphorylase kinase, gamma 2
Kinases & Inhibitors
PHK Kinase Family
Yes
Documents