Invitrogen™ PHKG1 Recombinant Human Protein DFS Item
PHKG1 is a serine/threonine protein kinase.
Manufacturer:  Invitrogen™ LSPV4034
 View more versions of this product
Catalog No. PV4034
Description
Description
PHKG1 is involved in glycogenolytic regulatory functions.
Specifications
Specifications
NP_006204
Frozen
72.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
PHK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
PHKG1
Baculovirus
Human
GST Tag
Documents