Thermo Scientific™ Phire Buffers
Choose from three formats of these buffers to optimize PCR using Thermo Scientific™ Phire™ Hot Start II DNA Polymerase.
$30.81 - $30.81
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity  
F525L Thermo Scientific™
F525L
PCR, High-Fidelity PCR, High Performance PCR 4 x 1.5mL Each for $30.81
F524L Thermo Scientific™
F524L
Phire DNA Polymerases. 4 x 1.5mL Each for $30.81
Specifications
Description
The reaction buffer for Thermo Scientific Phire Hot Start II DNA Polymerase is available in three formats: non-colored version with and without detergents or Green version containing a density reagent and two tracking dyes for direct loading of PCR products on gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.
The concentration of Mg2+ in the reaction buffers is optimized to work well for most amplicons. The 5X buffer contains 1.5 mM MgCl2 at final reaction concentration. A separate tube of DMSO is provided for further optimization.
Includes:
Detergent-free 5x Phire Reaction Buffer, 50mM MgCl2 solution, 100% DMSO
Recommended for:
PCR, High-Fidelity PCR, High Performance PCR