Please login to your online account to display your discounted pricing

PhiKan 083, Tocris™

p53 stabilizing agent

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material PhiKan 083
Assay Percent Range >99%
Molecular Formula C16H18N2·HCl
Formula Weight 274.79
Synonym 9-Ethyl-N-methyl-9H-carbazole-3-methanamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

432610

 
r & d systems
432610
10mg Each for $221.01

432650

 
r & d systems
432650
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 274.79
Quantity 10mg
CAS 880813-36-5
Molecular Formula C16H18N2·HCl
Chemical Name or Material PhiKan 083
Synonym 9-Ethyl-N-methyl-9H-carbazole-3-methanamine hydrochloride
Assay Percent Range >99%
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C