missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylurethane 98.0+%, TCI America™

Manufacturer:  TCI America P0165500G

 View more versions of this product

Catalog No. P0165500G



Specifications

Specifications

Phenylurethane
White-Yellow
500g
C9H11NO2
n-phenylurethane, phenylurethane, ethyl phenylcarbamate, phenylurethan, ethyl carbanilate, euphorin, keimstop, urethane, phenyl, carbanilic acid, ethyl ester, phenylethyl carbamate
CCOC(=O)NC1=CC=CC=C1
165.192
165.19
≥98.0% (GC)
237°C
52°C
101-99-5
MFCD00026806
LBKPGNUOUPTQKA-UHFFFAOYSA-N
ethyl N-phenylcarbamate
7591
Crystal-Powder at 20°C

Chemical Identifiers

101-99-5
165.192
LBKPGNUOUPTQKA-UHFFFAOYSA-N
7591
CCOC(=O)NC1=CC=CC=C1
C9H11NO2
MFCD00026806
n-phenylurethane, phenylurethane, ethyl phenylcarbamate, phenylurethan, ethyl carbanilate, euphorin, keimstop, urethane, phenyl, carbanilic acid, ethyl ester, phenylethyl carbamate
ethyl N-phenylcarbamate
Safety and Handling

Safety and Handling

RTECSNumber : FD8925000

TSCA : Yes

Documents