missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylurea, 97%

$51.11 - $142.53

Chemical Identifiers

CAS 64-10-8
Molecular Formula C7H8N2O
Linear Formula C6H5NHCONH2
Formula Weight 136.15
MDL Number MFCD00007944
Beilstein 1934615
Merck Index 14,7319
Chemical Name or Material Phenylurea
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0130636 SDS Alfa Aesar™
L0130636
500g Each for $142.53
AAL0130622 SDS Alfa Aesar™
L0130622
100g Each for $51.11
Specifications

Description

Applications
Phenyl urea is used in organic synthesis. It acts as an efficient ligand for palladium-catalyzed Heck and Suzuki reactions of aryl bromides and iodides.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

64-10-8
C6H5NHCONH2
MFCD00007944
14,7319
C7H8N2O
136.15
1934615
Phenylurea

Specifications

238°C
500g
C7H8N2O
136.15
1934615
Phenylurea
144° to 148°C
64-10-8
C6H5NHCONH2
MFCD00007944
14,7319
97%

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YU0650000

TSCA : TSCA