missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyltrimethylammonium Tribromide 97%, ACROS Organics™

Manufacturer:  Acros Organics AC160941000

 View more versions of this product

Catalog No. AC160941000



Description

Description

C-X bond formation reagent
Specifications

Specifications

Phenyltrimethylammonium tribromide
Plastic bottle
113.0°C to 117.0°C
97%
97%
C6H5N(CH3)3Br·Br2
01,855; 02,328; 04,386
Solubility in water: insoluble.
C[N+](C)(C)C1=CC=CC=C1.I[I-]I
516.931
375.92
97%
Authentic
Orange
100g
4207-56-1
C9H14I3N
MFCD00011789
mono n,n,n-trimethylbenzenaminium tribromide
RHXJPCQWYPXLEH-UHFFFAOYSA-N
trimethyl(phenyl)azanium;triiodide
21525000
Crystalline Powder

Chemical Identifiers

4207-56-1
516.931
RHXJPCQWYPXLEH-UHFFFAOYSA-N
21525000
C[N+](C)(C)C1=CC=CC=C1.I[I-]I
C9H14I3N
MFCD00011789
mono n,n,n-trimethylbenzenaminium tribromide
trimethyl(phenyl)azanium;triiodide
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-127-8

TSCA : TSCA

SDS Documents