missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyltrimethylammonium tribromide, 97%

Manufacturer:  Alfa Aesar™ A1532630

 View more versions of this product

Catalog No. AAA1532630



Description

Description

Applications
Phenyltrimethylammonium tribromide is used as brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.

Solubility
Insoluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents, reducing agents and bases.
Specifications

Specifications

Phenyltrimethylammonium tribromide
112°C to 117°C
4207-56-1
MFCD00011789
3919459
RHXJPCQWYPXLEH-UHFFFAOYSA-N
trimethyl(phenyl)azanium;triiodide
21525000
97%
Irritating
250g
C9H14I3N
UN3261
mono n,n,n-trimethylbenzenaminium tribromide
C[N+](C)(C)C1=CC=CC=C1.I[I-]I
516.931
375.93

Chemical Identifiers

4207-56-1
516.931
RHXJPCQWYPXLEH-UHFFFAOYSA-N
21525000
C[N+](C)(C)C1=CC=CC=C1.I[I-]I
C9H14I3N
MFCD00011789
mono n,n,n-trimethylbenzenaminium tribromide
trimethyl(phenyl)azanium;triiodide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Documents