missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyltrimethylammonium iodide, 99%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 160681000

 View more versions of this product

Catalog No. AC160681000



Description

Description

Phase transfer catalyst
Specifications

Specifications

Phenyltrimethylammonium iodide
Glass bottle
227.0°C
99%
98.5% min. (Argentometry)
C6H5N(CH3)3I
15, 7428
Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in chloroform
C[N+](C)(C)C1=CC=CC=C1.[I-]
263.11
263.11
99%
Authentic
Green to White
100g
98-04-4
C9H14IN
MFCD00011791
ammonium, phenyltrimethyl-, iodide
KKLAORVGAKUOPZ-UHFFFAOYSA-M
trimethyl(phenyl)azanium;iodide
94135
Slightly Crystalline Powder or Flakes

Chemical Identifiers

98-04-4
263.11
KKLAORVGAKUOPZ-UHFFFAOYSA-M
94135
C[N+](C)(C)C1=CC=CC=C1.[I-]
C9H14IN
MFCD00011791
ammonium, phenyltrimethyl-, iodide
trimethyl(phenyl)azanium;iodide
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-630-3

RTECSNumber : BT2450000

TSCA : TSCA

SDS Documents