missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyltrimethylammonium Iodide 99%, ACROS Organics™

$98.36 - $98.36

Chemical Identifiers

CAS 98-04-4
Assay Percent Range 99%
Molecular Formula C9H14IN
Linear Formula C6H5N(CH3)3I
Formula Weight 263.11
MDL Number MFCD00011791
Synonym (N, N, N)-Trimethylanilinium iodide
Chemical Name or Material Phenyltrimethylammonium iodide, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC160681000 SDS Thermo Fisher Scientific
160681000
Glass bottle 100g Each for $98.36
Specifications

Description

Phase transfer catalyst
Description & Specifications

Chemical Identifiers

98-04-4
C9H14IN
263.11
(N, N, N)-Trimethylanilinium iodide
≥98.5%
99%
C6H5N(CH3)3I
MFCD00011791
Phenyltrimethylammonium iodide, 99%

Specifications

263.11
White to slightly green
15, 7428
Authentic
98-04-4
C9H14IN
MFCD00011791
≥98.5%
Slightly Crystalline Powder or Flakes
100g
Glass bottle
(N, N, N)-Trimethylanilinium iodide
99%
C6H5N(CH3)3I
Phenyltrimethylammonium iodide, 99%

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 202-630-3

RTECSNumber : BT2450000

TSCA : TSCA