Please login to your online account to display your discounted pricing

Phenyltrimethylammonium Bromide 98%, ACROS Organics™

$101.60 - $101.60

Chemical Identifiers

Chemical Name or Material Phenyltrimethylammonium bromide
Name Note 98%
CAS 16056-11-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H14BrN
Linear Formula C6H5N(CH3)3Br
Formula Weight 216.12
MDL Number MFCD00011788
Synonym (N, N, N)-Trimethylanilinium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC159161000

  SDS
thermo fisher scientific
159161000
100g Each for $101.60
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder or Chunks
Formula Weight 216.12
Quantity 100g
Color White to blue
TSCA TSCA
RTECS Number BT2100000
EINECS Number 240-202-8
Infrared Spectrum Authentic
Synonym (N, N, N)-Trimethylanilinium bromide
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 16056-11-4
Health Hazard 1 HYGROSCOPIC
Linear Formula C6H5N(CH3)3Br
Name Note 98%
Chemical Name or Material Phenyltrimethylammonium bromide
Molecular Formula C9H14BrN
MDL Number MFCD00011788
Assay Percent Range 98%

Indicator, Phase transfer catalyst

Safety and Handling

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.