Please login to your online account to display your discounted pricing

Phenyltrimethoxysilane, 85%, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC370640010

 View more versions of this product

Catalog No. AC370640010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥80%
Formula Weight 198.29
Physical Form Liquid
Quantity 1L
Color Clear colorless
Density 1.0620g/mL
Beilstein 16, IV, 1556
Chemical Name or Material Phenyltrimethoxysilane
Packaging Glass bottle
Refractive Index 1.4730 to 1.4750 (20°C, 589nm)
Assay Percent Range 85%
Molecular Formula C9H14O3Si
Linear Formula C6H5Si(OCH3)3
MDL Number MFCD00025689
CAS 2996-92-1
Synonym Trimethoxysilylbenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Phenyltrimethoxysilane
Assay ≥80%
Assay Percent Range 85%
Molecular Formula C9H14O3Si
Linear Formula C6H5Si(OCH3)3
Formula Weight 198.29
MDL Number MFCD00025689
Synonym Trimethoxysilylbenzene
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents