missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyltrimethoxysilane, 85%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC370640500

 View more versions of this product

Catalog No. AC370640500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Phenyltrimethoxysilane
50°C
Glass bottle
1.062
Colorless
50mL
80% min. (GC)
C6H5Si(OCH3)3
16, IV, 1556
Solubility in water: soluble
CO[Si](OC)(OC)C1=CC=CC=C1
198.29
198.29
85%
1.0620g/mL
Authentic
1.4730 to 1.475
233°C
-25°C
2996-92-1
C9H14O3Si
MFCD00025689
benzene, trimethoxysilyl, phenyl trimethoxy silane, phenyl trimethoxysilane, phenyltri methoxy silane, phenyltrimethoxysilane, silane, phenyltrimethoxy, silane, trimethoxyphenyl, trimethoxy phenyl silane, trimethoxysilyl benzene, unii-21tqe746s9
ZNOCGWVLWPVKAO-UHFFFAOYSA-N
trimethoxy(phenyl)silane
18137
Liquid

Chemical Identifiers

2996-92-1
198.29
ZNOCGWVLWPVKAO-UHFFFAOYSA-N
18137
CO[Si](OC)(OC)C1=CC=CC=C1
C9H14O3Si
MFCD00025689
benzene, trimethoxysilyl, phenyl trimethoxy silane, phenyl trimethoxysilane, phenyltri methoxy silane, phenyltrimethoxysilane, silane, phenyltrimethoxy, silane, trimethoxyphenyl, trimethoxy phenyl silane, trimethoxysilyl benzene, unii-21tqe746s9
trimethoxy(phenyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if swallowed.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Warning

EINECSNumber : 221-066-9

SDS Documents