missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenyltriethoxysilane, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 174640010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Phenyltriethoxysilane | |
780-69-8 | |
235.0°C | |
Authentic | |
Glass bottle | |
1.4594 to 1.4614 | |
MFCD00009065 | |
16, 911 | |
phenyltriethoxysilane, triethoxy phenyl silane, silane, triethoxyphenyl, triethoxyfenylsilan, silane, phenyltriethoxy, benzeneorthosiliconic acid, triethyl ester, phenyl triethoxysilane, benzene, triethoxysilyl, triethoxyfenylsilan czech, unii-qi310t2x15 | |
JCVQKRGIASEUKR-UHFFFAOYSA-N | |
triethoxy(phenyl)silane | |
13075 | |
98% |
98% | |
0.9960g/mL | |
42°C | |
98% | |
C12H20O3Si | |
C6H5Si(OC2H5)3 | |
1kg | |
0.996 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
CCO[Si](OCC)(OCC)C1=CC=CC=C1 | |
240.37 | |
240.38 |
Chemical Identifiers
780-69-8 | |
240.37 | |
JCVQKRGIASEUKR-UHFFFAOYSA-N | |
13075 | |
CCO[Si](OCC)(OCC)C1=CC=CC=C1 |
C12H20O3Si | |
MFCD00009065 | |
phenyltriethoxysilane, triethoxy phenyl silane, silane, triethoxyphenyl, triethoxyfenylsilan, silane, phenyltriethoxy, benzeneorthosiliconic acid, triethyl ester, phenyl triethoxysilane, benzene, triethoxysilyl, triethoxyfenylsilan czech, unii-qi310t2x15 | |
triethoxy(phenyl)silane |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May cause damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 212-305-8
RTECSNumber : VV4900000
TSCA : TSCA
RUO â Research Use Only