Learn More
Phenyltriethoxysilane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0468430
Description
Phenyltriethoxysilane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenyltriethoxysilane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Hydrolyzes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
Specifications
235°C | |
780-69-8 | |
C12H20O3Si | |
MFCD00009065 | |
96°C (204°F) | |
Hydrolyzes in water. | |
CCO[Si](OCC)(OCC)C1=CC=CC=C1 | |
240.37 | |
240.38 | |
1.461 | |
0.996 |
Phenyltriethoxysilane | |
250 g | |
C6H5Si(OCH2CH3)3 | |
2940602 | |
phenyltriethoxysilane, triethoxy phenyl silane, silane, triethoxyphenyl, triethoxyfenylsilan, silane, phenyltriethoxy, benzeneorthosiliconic acid, triethyl ester, phenyl triethoxysilane, benzene, triethoxysilyl, triethoxyfenylsilan czech, unii-qi310t2x15 | |
JCVQKRGIASEUKR-UHFFFAOYSA-N | |
triethoxy(phenyl)silane | |
13075 | |
98% | |
Moisture sensitive |
Chemical Identifiers
780-69-8 | |
240.37 | |
JCVQKRGIASEUKR-UHFFFAOYSA-N | |
13075 | |
CCO[Si](OCC)(OCC)C1=CC=CC=C1 |
C12H20O3Si | |
MFCD00009065 | |
phenyltriethoxysilane, triethoxy phenyl silane, silane, triethoxyphenyl, triethoxyfenylsilan, silane, phenyltriethoxy, benzeneorthosiliconic acid, triethyl ester, phenyl triethoxysilane, benzene, triethoxysilyl, triethoxyfenylsilan czech, unii-qi310t2x15 | |
triethoxy(phenyl)silane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P260-P280-P303+P361+P353-P314-P370+P378q-P501c
H226-H373
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 212-305-8
RTECSNumber : VV4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only