Learn More
Phenyltrichlorosilane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1671322
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
201°C | |
-127°C | |
100 g | |
(C6H5)SiCl3 | |
UN1804 | |
91°C (195°F) | |
ORVMIVQULIKXCP-UHFFFAOYSA-N | |
trichloro(phenyl)silane | |
7372 | |
97% | |
1.523 | |
1.324 |
Phenyltrichlorosilane | |
98-13-5 | |
C6H5Cl3Si | |
MFCD00000479 | |
508730 | |
trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane | |
C1=CC=C(C=C1)[Si](Cl)(Cl)Cl | |
211.541 | |
211.55 | |
Odorless | |
Moisture sensitive |
Chemical Identifiers
98-13-5 | |
211.541 | |
ORVMIVQULIKXCP-UHFFFAOYSA-N | |
7372 | |
C1=CC=C(C=C1)[Si](Cl)(Cl)Cl |
C6H5Cl3Si | |
MFCD00000479 | |
trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane | |
trichloro(phenyl)silane |
Safety and Handling
GHS H Statement
H330-H314-H318-H227-H312
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful in contact with skin.
P210-P235-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H311-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHENYLTRICHLOROSILANE
EINECSNumber : 202-640-8
RTECSNumber : VV6650000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only