Learn More
Phenyltrichlorosilane, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 131000100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Phenyltrichlorosilane | |
98-13-5 | |
201°C | |
Authentic | |
Glass bottle | |
1.5220 to 1.524 | |
MFCD00000479 | |
16, 911 | |
trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane | |
ORVMIVQULIKXCP-UHFFFAOYSA-N | |
trichloro(phenyl)silane | |
7372 | |
95% |
95% | |
1.3210g/mL | |
91°C | |
94% min. (GC) | |
C6H5Cl3Si | |
C6H5SiCl3 | |
10mL | |
1.321 | |
Solubility in water: reacts. Other solubilities: soluble in benzene,ehter and perchloroethylene | |
C1=CC=C(C=C1)[Si](Cl)(Cl)Cl | |
211.55 | |
211.55 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Reacts violently with water.
Corrosive to the respiratory tract.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 202-640-8
RTECSNumber : VV6650000
TSCA : TSCA
RUO â Research Use Only