missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyltin trichloride

Manufacturer:  Alfa Aesar™ 7115706

 View more versions of this product

Catalog No. AA7115706



Specifications

Specifications

Phenyltin trichloride
5g
1.84g/mL
C6H5Cl3Sn
UN2788
UBOGEXSQACVGEC-UHFFFAOYSA-K
trichloro(phenyl)stannane
14298
Liquid
1124-19-2
Pungent
Moisture Sensitive
MFCD00000513
phenyltin trichloride, phenyltrichlorotin, stannane, trichlorophenyl, trichlorophenyltin, trichloro phenyl stannane, phenyltrichlorostannane, ccris 6323, phsncl3, phenyl tin trichloride, monophenyltin trichloride
C1=CC=C(C=C1)[Sn](Cl)(Cl)Cl
302.166
302.18

Chemical Identifiers

1124-19-2
302.166
UBOGEXSQACVGEC-UHFFFAOYSA-K
14298
C1=CC=C(C=C1)[Sn](Cl)(Cl)Cl
C6H5Cl3Sn
MFCD00000513
phenyltin trichloride, phenyltrichlorotin, stannane, trichlorophenyl, trichlorophenyltin, trichloro phenyl stannane, phenyltrichlorostannane, ccris 6323, phsncl3, phenyl tin trichloride, monophenyltin trichloride
trichloro(phenyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

SDS Documents