Please login to your online account to display your discounted pricing

Phenylthioacetonitrile, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Phenylthioacetonitrile
CAS 5219-61-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H7NS
Formula Weight 149.22
MDL Number MFCD00019831
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC355330050

  SDS
thermo fisher scientific
35533-0050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 149.22
Quantity 5g
Color Clear colorless to light yellow
EINECS Number 226-013-3
RTECS Number AM1900000
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Infrared Spectrum Authentic
Molecular Formula C8H7NS
Chemical Name or Material Phenylthioacetonitrile
Refractive Index 1.5790 to 1.5810 (20°C, 589nm)
MDL Number MFCD00019831
CAS 5219-61-4
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.