Please login to your online account to display your discounted pricing

Alfa Aesar™ (Phenylthio)trimethylsilane, 97%

Chemical Identifiers

Chemical Name or Material (Phenylthio)trimethylsilane
CAS 4551-15-9
Molecular Formula C6H5SSi(CH3)3
Formula Weight 182.36
MDL Number MFCD00008272
UN Number UN1993
Beilstein 1905944
Synonym Phenyl trimethylsilyl sulfide; S-Trimethylsilylthiophenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0631906

  SDS
alfa aesar™
L0631906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0631914

  SDS
alfa aesar™
L0631914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.36
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material (Phenylthio)trimethylsilane
CAS 4551-15-9
Health Hazard 1 Warning
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA TSCA
MDL Number MFCD00008272
Molecular Formula C6H5SSi(CH3)3
Odor Stench
Assay 97%
Flash Point 30°C (86°F)
Beilstein 1905944
UN Number UN1993
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Boiling Point 73° to 75°C (10mmHg)
Synonym Phenyl trimethylsilyl sulfide; S-Trimethylsilylthiophenol

Applications
(Phenylthio)trimethylsilane is used to produce 1-phenyl-1,2-bis-phenylsulfanyl-ethane at the temperature of 40°C.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with moisture, oxidizing agents, Strong acids. Protect from humidity and water. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.