Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylsulfonylacetonitrile, 98%

Chemical Identifiers

Chemical Name or Material Phenylsulfonylacetonitrile
CAS 7605-28-9
Molecular Formula C8H7NO2S
Formula Weight 181.22
MDL Number MFCD00007550
UN Number UN3439
Beilstein 640716
Synonym Benzenesulfonylacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1542718

  SDS
alfa aesar™
A1542718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1542730

  SDS
alfa aesar™
A1542730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.22
Chemical Name or Material Phenylsulfonylacetonitrile
Assay 98%
Molecular Formula C8H7NO2S
MDL Number MFCD00007550
CAS 7605-28-9
UN Number UN3439
Beilstein 640716
Synonym Benzenesulfonylacetonitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 110° to 114°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Quantity 50g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.