Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylsulfonylacetone, 97%

$47.72 - $769.64

Chemical Identifiers

Chemical Name or Material Phenylsulfonylacetone
CAS 5000-44-2
Molecular Formula C9H10O3S
Formula Weight 198.24
MDL Number MFCD00047794
Beilstein 640928
Synonym Benzenesulfonylacetone; 1-Phenylsulfonyl-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1408503

 
alfa aesar™
A1408503
1g Each for $47.72

AAA1408514

 
alfa aesar™
A1408514
25g Each for $769.64
Description & Specifications

Specifications

Formula Weight 198.24
Chemical Name or Material Phenylsulfonylacetone
Assay 97%
Molecular Formula C9H10O3S
MDL Number MFCD00047794
CAS 5000-44-2
Beilstein 640928
Synonym Benzenesulfonylacetone; 1-Phenylsulfonyl-2-propanone
Melting Point 54° to 58°C
Quantity 1g