Please login to your online account to display your discounted pricing

Alfa Aesar™ (Phenylsulfonyl)acetamide, 98+%

$38.12 - $132.45

Chemical Identifiers

Chemical Name or Material (Phenylsulfonyl)acetamide
CAS 35008-50-5
Molecular Formula C8H9NO3S
Formula Weight 199.23
MDL Number MFCD00041320
Beilstein 2101624
Synonym (Benzenesulfonyl)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1274106

  SDS
alfa aesar™
L1274106
5g Each for $38.12

AAL1274114

  SDS
alfa aesar™
L1274114
25g Each for $132.45
Description & Specifications

Specifications

Formula Weight 199.23
Chemical Name or Material (Phenylsulfonyl)acetamide
Assay ≥98%
Molecular Formula C8H9NO3S
MDL Number MFCD00041320
CAS 35008-50-5
Synonym (Benzenesulfonyl)acetamide
Beilstein 2101624
Melting Point 154° to 158°C
Quantity 5g