Alfa Aesar™ (Phenylsulfonyl)acetamide, 98+%
Manufacturer:  Alfa Aesar™ L1274106
 View more versions of this product
Catalog No. AAL1274106
Specifications
Specifications
(Phenylsulfonyl)acetamide
5g
C8H9NO3S
2101624
LKYNWTGTRVZBPA-UHFFFAOYSA-N
2-(benzenesulfonyl)acetamide
1810305
≥98%
154°C to 158°C
35008-50-5
MFCD00041320
2-benzenesulfonyl acetamide, 2-phenylsulfonyl acetamide, phenylsulfonyl acetamide, phenylsulfonylacetamide, acmc-20aplb, enamine_000243, phenylsulphonyl acetamide, alpha-phenylsulfonylacetamide, 2-phenylsulfonyl ethanamide, acetamide, 2-phenylsulfonyl
C1=CC=C(C=C1)S(=O)(=O)CC(=O)N
199.224
199.23
Chemical Identifiers
35008-50-5
199.224
LKYNWTGTRVZBPA-UHFFFAOYSA-N
1810305
C1=CC=C(C=C1)S(=O)(=O)CC(=O)N
C8H9NO3S
MFCD00041320
2-benzenesulfonyl acetamide, 2-phenylsulfonyl acetamide, phenylsulfonyl acetamide, phenylsulfonylacetamide, acmc-20aplb, enamine_000243, phenylsulphonyl acetamide, alpha-phenylsulfonylacetamide, 2-phenylsulfonyl ethanamide, acetamide, 2-phenylsulfonyl
2-(benzenesulfonyl)acetamide
Documents