missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenylsuccinic Anhydride 98.0+%, TCI America™
Supplier: TCI America P225725G
Specifications
Phenylsuccinic Anhydride | |
54°C | |
C10H8O3 | |
25 g | |
HDFKMLFDDYWABF-UHFFFAOYNA-N | |
3-phenyloxolane-2,5-dione | |
136911 | |
≥98.0% (GC,T) |
1131-15-3 | |
195°C | |
MFCD00192395 | |
phenylsuccinic anhydride, 3-phenyldihydrofuran-2,5-dione, monophenyl succinic anhydride, 2,5-furandione, dihydro-3-phenyl, acmc-20ajwd, acmc-20mmqc, phenylsuccinicanhydride, 2,5-furandione, dihydro-3-phenyl-, 3r, phenyl succinic anhydride | |
O=C1CC(C(=O)O1)C1=CC=CC=C1 | |
176.17 | |
176.17 | |
Crystalline Powder |
Chemical Identifiers
1131-15-3 | |
176.17 | |
HDFKMLFDDYWABF-UHFFFAOYNA-N | |
136911 | |
O=C1CC(C(=O)O1)C1=CC=CC=C1 |
C10H8O3 | |
MFCD00192395 | |
phenylsuccinic anhydride, 3-phenyldihydrofuran-2,5-dione, monophenyl succinic anhydride, 2,5-furandione, dihydro-3-phenyl, acmc-20ajwd, acmc-20mmqc, phenylsuccinicanhydride, 2,5-furandione, dihydro-3-phenyl-, 3r, phenyl succinic anhydride | |
3-phenyloxolane-2,5-dione |
Safety and Handling
TSCA : No