Please login to your online account to display your discounted pricing

Phenylsuccinic anhydride, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC428510050

 View more versions of this product

Catalog No. AC428510050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 176.17
Quantity 5g
Packaging Glass bottle
Molecular Formula C10H8O3
CAS 1131-15-3
Chemical Name or Material Phenylsuccinic anhydride
MDL Number MFCD00192395
Assay Percent Range 99%

Specifications

Chemical Name or Material Phenylsuccinic anhydride
Assay Percent Range 99%
Molecular Formula C10H8O3
Formula Weight 176.17
MDL Number MFCD00192395