missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenylsuccinic anhydride, 99%, Thermo Scientific™
$59.95 - $189.77
Chemical Identifiers
CAS | 1131-15-3 |
---|---|
Molecular Formula | C10H8O3 |
Molecular Weight (g/mol) | 176.17 |
MDL Number | MFCD00192395 |
InChI Key | HDFKMLFDDYWABF-UHFFFAOYNA-N |
Synonym | 2,5-furandione, dihydro-3-phenyl, 2,5-furandione, dihydro-3-phenyl-, 3r, 3-phenyldihydrofuran-2,5-dione, acmc-20ajwd, acmc-20mmqc, monophenyl succinic anhydride, phenyl succinic anhydride, phenylsuccinic anhydride, phenylsuccinicanhydride |
PubChem CID | 136911 |
IUPAC Name | 3-phenyloxolane-2,5-dione |
SMILES | O=C1CC(C(=O)O1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC428510050
|
Thermo Fisher Scientific
AC428510050 |
5g | Glass bottle |
Each for $59.95
|
|
AC428510250
|
Thermo Fisher Scientific
AC428510250 |
25g | Glass bottle |
Each for $189.77
|
|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1131-15-3 | |
176.17 | |
HDFKMLFDDYWABF-UHFFFAOYNA-N | |
136911 | |
O=C1CC(C(=O)O1)C1=CC=CC=C1 |
C10H8O3 | |
MFCD00192395 | |
2,5-furandione, dihydro-3-phenyl, 2,5-furandione, dihydro-3-phenyl-, 3r, 3-phenyldihydrofuran-2,5-dione, acmc-20ajwd, acmc-20mmqc, monophenyl succinic anhydride, phenyl succinic anhydride, phenylsuccinic anhydride, phenylsuccinicanhydride | |
3-phenyloxolane-2,5-dione |
Specifications
1131-15-3 | |
191°C to 192°C (12.0mmHg) | |
C10H8O3 | |
Glass bottle | |
5g | |
O=C1CC(C(=O)O1)C1=CC=CC=C1 | |
176.17 | |
176.17 | |
Phenylsuccinic anhydride |
53°C to 55°C | |
98.5% min. (GC) | |
MFCD00192395 | |
2,5-furandione, dihydro-3-phenyl, 2,5-furandione, dihydro-3-phenyl-, 3r, 3-phenyldihydrofuran-2,5-dione, acmc-20ajwd, acmc-20mmqc, monophenyl succinic anhydride, phenyl succinic anhydride, phenylsuccinic anhydride, phenylsuccinicanhydride | |
HDFKMLFDDYWABF-UHFFFAOYNA-N | |
3-phenyloxolane-2,5-dione | |
136911 | |
99% |
RUO – Research Use Only