Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylsuccinic anhydride, 99%

$64.50 - $669.58

Chemical Identifiers

Chemical Name or Material Phenylsuccinic anhydride
CAS 1131-15-3
Molecular Formula C10H8O3
Formula Weight 176.17
MDL Number MFCD00192395
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2441806

  SDS
alfa aesar™
B2441806
5g Each for $64.50

AAB2441814

  SDS
alfa aesar™
B2441814
25g Each for $204.73

AAB2441822

  SDS
alfa aesar™
B2441822
100g Each for $669.58
Description & Specifications

Specifications

Formula Weight 176.17
Chemical Name or Material Phenylsuccinic anhydride
Assay 99%
Molecular Formula C10H8O3
MDL Number MFCD00192395
CAS 1131-15-3
Boiling Point 191° to 192°C (12mmHg)
Melting Point 53° to 55°C
Quantity 5g