missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-Phenylsuccinic acid, 98%

Manufacturer:  Alfa Aesar™ A1197114

 View more versions of this product

Catalog No. AAA1197114


 

Specifications

Specifications

(±)-Phenylsuccinic acid
635-51-8
C10H10O4
1875051
C1=CC=C(C=C1)C(CC(=O)O)C(=O)O
194.186
194.19
25g
98%
MFCD00004256
LVFFZQQWIZURIO-UHFFFAOYSA-N
2-phenylbutanedioic acid
95459
167°C to 169°C

Chemical Identifiers

635-51-8
194.186
LVFFZQQWIZURIO-UHFFFAOYSA-N
C10H10O4
MFCD00004256
95459
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents