missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(+/-)-Phenylsuccinic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1197122
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(±)-Phenylsuccinic acid | |
167°C to 169°C | |
MFCD00004256 | |
1875051 | |
LVFFZQQWIZURIO-UHFFFAOYSA-N | |
2-phenylbutanedioic acid | |
95459 | |
98% |
635-51-8 | |
C10H10O4 | |
100 g | |
phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl | |
C1=CC=C(C=C1)C(CC(=O)O)C(=O)O | |
194.186 | |
194.19 |
Chemical Identifiers
635-51-8 | |
194.186 | |
LVFFZQQWIZURIO-UHFFFAOYSA-N | |
95459 | |
C1=CC=C(C=C1)C(CC(=O)O)C(=O)O |
C10H10O4 | |
MFCD00004256 | |
phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl | |
2-phenylbutanedioic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-238-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only