missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylsilane, 97+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC291691000

 View more versions of this product

Catalog No. AC291691000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Reduction reagent
Specifications

Specifications

Phenylsilane
7°C
Glass bottle
0.87
Colorless
694-53-1
C6H8Si
03,1198
benzene, silyl, fenylsilan, fenylsilan czech, phenylsilane, phenylsilane, silyl, silane, phenyl, silane, phenyl-,, silylbenzene
XJWOWXZSFTXJEX-UHFFFAOYSA-N
phenylsilicon
6327628
Liquid
0.8700g/mL
Authentic
1.5100 to 1.5120
120.0°C
100mL
97% min. (GC)
C6H5SiH3
05,523; 17,284
Solubility in water: reacts. Other solubilities: soluble in most common organic solvents
C1=CC=C(C=C1)[Si]
108.22
108.22
97+%

Chemical Identifiers

694-53-1
108.22
benzene, silyl, fenylsilan, fenylsilan czech, phenylsilane, phenylsilane, silyl, silane, phenyl, silane, phenyl-,, silylbenzene
phenylsilicon
C6H8Si
XJWOWXZSFTXJEX-UHFFFAOYSA-N
6327628
C1=CC=C(C=C1)[Si]
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
In contact with water release

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

EINECSNumber : 211-772-5

SDS Documents