Learn More
Phenylselenol, 98%, ACROS Organics™
Chemical Identifiers
CAS | 645-96-5 |
---|---|
Molecular Formula | C6H6Se |
Molecular Weight (g/mol) | 157 |
MDL Number | MFCD00000001 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC123520010
|
Thermo Fisher Scientific
AC123520010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC123520100
|
Thermo Fisher Scientific
123520100 |
10g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
645-96-5 | |
C6H6Se | |
157 | |
MFCD00000001 |
Specifications
70°C | |
Glass bottle | |
183.0°C | |
1g | |
97.5% min. (GC) | |
C6H5SeH | |
157 | |
Liquid | |
Phenylselenol, 98% |
Authentic | |
1.6150 to 1.6180 | |
Yellow | |
645-96-5 | |
C6H6Se | |
MFCD00000001 | |
157 | |
98% |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 211-457-2
TSCA : TSCA