Learn More
Phenylselenol, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 123520100
Specifications
Phenylselenol | |
Authentic | |
1.6150 to 1.6180 | |
Yellow | |
98% | |
97.5% min. (GC) | |
C6H5SeH | |
157 | |
Liquid |
70°C | |
Glass bottle | |
183.0°C | |
10g | |
645-96-5 | |
C6H6Se | |
MFCD00000001 | |
157 | |
98% |
Chemical Identifiers
645-96-5 | |
C6H6Se | |
157 | |
MFCD00000001 |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 211-457-2
TSCA : TSCA