Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylselenol

Chemical Identifiers

Chemical Name or Material Phenylselenol
CAS 645-96-5
Formula Weight 157.03g/mol
MDL Number MFCD00000001
UN Number UN3440
Synonym Benzeneselenol; Selenophenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1572003

 
alfa aesar™
15720-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1572006

 
alfa aesar™
15720-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA15720MD

 
alfa aesar™
15720-MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Boiling Point 71° to 72°C (18mmHg)
Quantity 1g
Flash Point 70° (158°F)
Chemical Name or Material Phenylselenol
CAS 645-96-5
Physical Form Liquid
Formula Weight 157.03g/mol
MDL Number MFCD00000001
UN Number UN3440
Synonym Benzeneselenol; Selenophenol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331-H373-H227
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P261-P301+P310-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, LIQUID, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P260-P261-P301+P310-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H373-H227
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.