missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenylpyruvic acid, sodium salt monohydrate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 189730250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Phenylpyruvic acid, sodium salt monohydrate | |
122049-54-1 | |
98% | |
C9H7O3 | |
MFCD00149109 | |
10, I, 325 | |
Solubility in water: soluble in water | |
[O-]C(=O)C(=O)CC1=CC=CC=C1 | |
163.15 | |
204.16 |
98% | |
Authentic | |
Glass bottle | |
C6H5CH2COCO2Na·H2O | |
25g | |
sodium phenylpyruvate monohydrate, sodium 2-oxo-3-phenylpropanoate hydrate, phenylpyruvic acid sodium salt monohydrate, phenylpyruvic acid, sodium salt, sodium hydrate ?-phenylpyruvate, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, acmc-2099lh, phenylpyruvic acid sodium hydrate, beta-phenylpyruvic acid, sodium salt | |
BTNMPGBKDVTSJY-UHFFFAOYSA-M | |
sodium;2-oxo-3-phenylpropanoate;hydrate | |
23666336 | |
98% |
Chemical Identifiers
122049-54-1 | |
163.15 | |
BTNMPGBKDVTSJY-UHFFFAOYSA-M | |
23666336 | |
[O-]C(=O)C(=O)CC1=CC=CC=C1 |
C9H7O3 | |
MFCD00149109 | |
sodium phenylpyruvate monohydrate, sodium 2-oxo-3-phenylpropanoate hydrate, phenylpyruvic acid sodium salt monohydrate, phenylpyruvic acid, sodium salt, sodium hydrate ?-phenylpyruvate, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, acmc-2099lh, phenylpyruvic acid sodium hydrate, beta-phenylpyruvic acid, sodium salt | |
sodium;2-oxo-3-phenylpropanoate;hydrate |
Safety and Handling
EINECSNumber : 204-053-2
RUO â Research Use Only