Learn More
Phenylphosphonic dichloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 130800010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Phenylphosphonic dichloride | |
824-72-6 | |
258.0°C | |
Authentic | |
Glass bottle | |
1.5570 to 1.5610 | |
MFCD00002070 | |
16, 804 | |
1.39 | |
Solubility in water: reacts | |
C1=CC=C(C=C1)P(=O)(Cl)Cl | |
194.98 | |
194.98 |
97% | |
1.3900g/mL | |
204°C | |
96% min. (Argentometry) | |
C6H5Cl2OP | |
C6H5P(O)Cl2 | |
1L | |
01,846; 16,284 | |
phenylphosphonic dichloride, phosphonic dichloride, phenyl, benzenephosphonic dichloride, phenylphosphonyl dichloride, dichlorophenylphosphine oxide, phenyldichlorophosphine oxide, benzenephosphonyl chloride, phenylphosphonic acid dichloride, phenylphosphonodichloridic acid, phosphonic dichloride, p-phenyl | |
IBDMRHDXAQZJAP-UHFFFAOYSA-N | |
dichlorophosphorylbenzene | |
69990 | |
97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May be corrosive to metals.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 212-534-3
TSCA : TSCA
RUO â Research Use Only