Please login to your online account to display your discounted pricing

Phenylphosphonic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  130790050

 View more versions of this product

Catalog No. AC130790050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 158.09
Physical Form Crystalline Powder
Assay ≥97.5%
Quantity 5g
Color White to off-white
TSCA TSCA
RTECS Number TA0350000
EINECS Number 216-388-1
Melting Point 158° to 165°C
Synonym Benzenephosphonic acid
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass Bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Phenylphosphonic acid
MDL Number MFCD00002136
Name Note 98%
CAS 1571-33-1
Molecular Formula C6H7O3P
Linear Formula C6H5P(O)(OH)2
Assay Percent Range 98%

Specifications

Chemical Name or Material Phenylphosphonic acid
Name Note 98%
CAS 1571-33-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H7O3P
Linear Formula C6H5P(O)(OH)2
Formula Weight 158.09
MDL Number MFCD00002136
Synonym Benzenephosphonic acid
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.

SDS & Safety Documents