Please login to your online account to display your discounted pricing

Phenylphosphinic Acid 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Phenylphosphinic acid
Name Note 99%
CAS 1779-48-2
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H7O2P
Linear Formula C6H5P(O)(OH)H
Formula Weight 142.09
MDL Number MFCD00002131
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC130780050

  SDS
thermo fisher scientific
130780050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Solid
Formula Weight 142.09
Quantity 5g
Color White
EINECS Number 217-217-3
TSCA TSCA
RTECS Number SZ5462500
Melting Point 82° to 87°C
Beilstein 16, 791
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Chemical Name or Material Phenylphosphinic acid
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H7O2P
Linear Formula C6H5P(O)(OH)H
MDL Number MFCD00002131
CAS 1779-48-2
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Harmful if swallowed.