Please login to your online account to display your discounted pricing

Phenylphosphinic Acid 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material Phenylphosphinic acid
Name Note 97%
CAS 1779-48-2
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H7O2P
Linear Formula C6H5P(O)(OH)H
Formula Weight 142.09
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC417705000

  SDS
thermo fisher scientific
417705000
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 142.09
Physical Form Crystals or Crystalline Powder
Quantity 500g
Color White
Beilstein 16, 791
RTECS Number SZ5462500
EINECS Number 217-217-3
TSCA TSCA
Melting Point 82° to 87°C
Linear Formula C6H5P(O)(OH)H
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Infrared Spectrum Authentic
Assay Percent Range 97%
CAS 1779-48-2
Chemical Name or Material Phenylphosphinic acid
Name Note 97%
Molecular Formula C6H7O2P
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Harmful if swallowed.