Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylphosphinic acid, 97%

Chemical Identifiers

Chemical Name or Material Phenylphosphinic acid
CAS 1779-48-2
Formula Weight 142.09g/mol
MDL Number MFCD00002131
UN Number UN3261
Synonym Benzenephosphinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3009818

 
alfa aesar™
30098-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50g
Melting Point 83° to 85°C
Quantity 50g
Chemical Name or Material Phenylphosphinic acid
CAS 1779-48-2
Physical Form Crystalline
Assay 97%
Formula Weight 142.09g/mol
MDL Number MFCD00002131
UN Number UN3261
Synonym Benzenephosphinic acid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
I
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
I

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.