Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylphosphine

$19.78 - $19.78

Chemical Identifiers

Chemical Name or Material Phenylphosphine
CAS 638-21-1
Formula Weight 110.1
MDL Number MFCD00002094
UN Number UN2845
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3015503

 
alfa aesar™
3015503
1g Each for $19.78
Description & Specifications

Specifications

Formula Weight 110.1
Health Hazard 3 GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Health Hazard 2 GHS H Statement
H250-H252-H332-H315-H319-H335-H227
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Chemical Name or Material Phenylphosphine
Physical Form Liquid
MDL Number MFCD00002094
UN Number UN2845
Density 1.00g/mL
Odor Stench
Sensitivity Air & Moisture Sensitive
Quantity 1g
CAS 638-21-1
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
Boiling Point 160°C
Safety and Handling

Danger

GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.

GHS H Statement
H250-H252-H332-H315-H319-H335-H227
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.