missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylmercury(II) chloride, 96%, Hg 63.5%

$44.78 - $44.78

Chemical Identifiers

CAS 100-56-1
Formula Weight 313.18
MDL Number MFCD00000463
UN Number UN2026
Merck Index 14,7301
Synonym Phenylmercuric chloride
Chemical Name or Material Phenylmercury(II) chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3711309 SDS Alfa Aesar™
3711309
10g Each for $44.78
AA3711318 SDS Alfa Aesar™
3711318
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

100-56-1
MFCD00000463
14,7301
Phenylmercury(II) chloride
313.18
UN2026
Phenylmercuric chloride

Specifications

Hg 63.5%
100-56-1
313.18
UN2026
Phenylmercuric chloride
White
Phenylmercury(II) chloride
10g
Powder
MFCD00000463
14,7301
Soluble in benzene, ether, pyridine; Very slightly soluble in water
246° to 250°C
96%

Safety and Handling

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P280h-P273-P309+P311-P302+P352-P420a-P501a
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store away from foodstuffs.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLMERCURIC COMPOUNDS, N.O.S.

RTECSNumber : OW1400000