missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylmaleic Anhydride, TCI America™

Manufacturer:  TCI America P20965G

 View more versions of this product

Catalog No. P20965G



Specifications

Specifications

Phenylmaleic Anhydride
122°C
36122-35-7
MFCD00015472
QZYCWJVSPFQUQC-UHFFFAOYSA-N
3-phenylfuran-2,5-dione
99174
Crystal-Powder at 20°C
White-Yellow
5g
C10H6O3
phenylmaleic anhydride, 3-phenyl-furan-2,5-dione, 3-phenyl-2,5-furandione, 3-phenyl-2,5-dihydrofuran-2,5-dione, 2,5-furandione, 3-phenyl, phenylmaleicanhydride, phenylmalecanhydrde, acmc-20ajwi, phenyl maleic anhydride, phenyl-maleic anhydride
C1=CC=C(C=C1)C2=CC(=O)OC2=O
174.155
174.16
≥98.0% (GC,T)

Chemical Identifiers

36122-35-7
174.155
QZYCWJVSPFQUQC-UHFFFAOYSA-N
99174
C1=CC=C(C=C1)C2=CC(=O)OC2=O
C10H6O3
MFCD00015472
phenylmaleic anhydride, 3-phenyl-furan-2,5-dione, 3-phenyl-2,5-furandione, 3-phenyl-2,5-dihydrofuran-2,5-dione, 2,5-furandione, 3-phenyl, phenylmaleicanhydride, phenylmalecanhydrde, acmc-20ajwi, phenyl maleic anhydride, phenyl-maleic anhydride
3-phenylfuran-2,5-dione
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3109

TSCA : No

Documents