Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylmaleic anhydride, 97%

$45.41 - $157.00

Chemical Identifiers

Chemical Name or Material Phenylmaleic anhydride
CAS 36122-35-7
Molecular Formula C10H6O3
Formula Weight 174.16
MDL Number MFCD00015472
Beilstein 127301
Synonym 3-Phenyl-2,5-furandione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0345303

 
alfa aesar™
L0345303
1g Each for $45.41

AAL0345306

 
alfa aesar™
L0345306
5g Each for $157.00
Description & Specifications

Specifications

Formula Weight 174.16
Chemical Name or Material Phenylmaleic anhydride
CAS 36122-35-7
Molecular Formula C10H6O3
MDL Number MFCD00015472
Beilstein 127301
Synonym 3-Phenyl-2,5-furandione
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 118° to 119°C
Assay 97%
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.