Learn More
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 445968000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Phenylmagnesium bromide | |
−1°C | |
C6H5BrMg | |
MFCD00000038 | |
phenylmagnesium bromide, grignard reagent, magnesium, bromophenyl, phmgbr, phmgbr, bromo phenyl magnesium, bromo phenyl magnesium, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, bromomagnesiobenzene, phenyhnagnesium bromide | |
NIXOIRLDFIPNLJ-UHFFFAOYSA-M | |
magnesium;benzene;bromide | |
66852 |
100-58-3,5614-37-9 | |
AcroSeal™ Glass bottle | |
C6H5MgBr | |
800mL | |
Solubility in water: reacts | |
[Mg++].[Br-].c1cc[cH-]cc1 | |
181.32 | |
181.31 |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 202-867-2
RUO â Research Use Only