missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylmagnesium bromide, 3M in ether

Grignard reagent

Manufacturer:  Alfa Aesar™ 87326AE

 View more versions of this product

Catalog No. AA87326AE



Specifications

Specifications

Phenylmagnesium bromide
Ether-like
Yellow to Brown
100-58-3
MFCD00000038
phenylmagnesium bromide, bromophenylmagnesium, magnesium, bromophenyl, phmgbr, phenylmagnesiumbromide, bromo phenyl magnesium, phenyl magnesium bromide, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide
NIXOIRLDFIPNLJ-UHFFFAOYSA-M
magnesium;benzene;bromide
66852
Liquid
1.14g/mL
Light, Air & Moisture Sensitive
100mL
C6H5BrMg
UN3399
3M in ether
C1=CC=[C-]C=C1.[Mg+2].[Br-]
181.315
181.31

Chemical Identifiers

100-58-3
181.315
NIXOIRLDFIPNLJ-UHFFFAOYSA-M
66852
C1=CC=[C-]C=C1.[Mg+2].[Br-]
C6H5BrMg
MFCD00000038
phenylmagnesium bromide, bromophenylmagnesium, magnesium, bromophenyl, phmgbr, phenylmagnesiumbromide, bromo phenyl magnesium, phenyl magnesium bromide, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide
magnesium;benzene;bromide
Safety and Handling

Safety and Handling

GHS H Statement
H224-H261-H314-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

TSCA : TSCA

SDS Documents