Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylhydroquinone diacetate, 98%

$34.64 - $111.59

Chemical Identifiers

Chemical Name or Material Phenylhydroquinone diacetate
CAS 58244-28-3
Molecular Formula C16H14O4
Formula Weight 270.29
MDL Number MFCD00145106
Beilstein 2383464
Synonym Biphenyl-2,5-diol diacetate; 2,5-Diacetoxybiphenyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2143309

 
alfa aesar™
B2143309
10g Each for $34.64

AAB2143318

 
alfa aesar™
B2143318
50g Each for $111.59
Description & Specifications

Specifications

Formula Weight 270.29
Chemical Name or Material Phenylhydroquinone diacetate
Assay 98%
Molecular Formula C16H14O4
MDL Number MFCD00145106
CAS 58244-28-3
Beilstein 2383464
Synonym Biphenyl-2,5-diol diacetate; 2,5-Diacetoxybiphenyl
Melting Point 67° to 70°C
RTECS Number DV4795000
Quantity 10g